siRNA Details
Gene Symbol: Mctp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024703.1 | NP_001019874.1 | 2 | 422 | ||
GenBank mRNA | AK156983.1 | - | 2 | 436 | ||
BC086658.1 | - | 2 | 422 | 30546156 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
