siRNA Details
Gene Symbol: Mctp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024703.1 | NP_001019874.1 | 22 | 2712 | ||
GenBank mRNA | BC086658.1 | - | 22 | 2712 | 30546156 |

More Information
Other Species Targeted

Gene Ontology Categories:
