siRNA Details
Gene Symbol: Rgma | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177740.5 | NP_808408.2 | 3 | 846 | ||
GenBank mRNA | AJ557513.1 | - | 2 | 600 | ||
AK082560.1 | - | 2 | 743 | |||
AK145224.1 | - | 2 | 589 | |||
BC023870.1 | - | 2 | 654 | 5353958 | ||
BC059072.1 | - | 3 | 846 | 6826181 |

More Information
Other Species Targeted

Gene Ontology Categories:
