siRNA Details
Gene Symbol: Ubqln3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198623.2 | NP_941025.1 | 2 | 781 | ||
GenBank mRNA | AK077072.1 | - | 2 | 783 | ||
BC100419.1 | - | 2 | 758 | 30918401 | ||
BC150726.1 | - | 1 | 708 | 9087637 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
