siRNA Details
Gene Symbol: E030002O03Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172905.2 | NP_766493.1 | 4 | 537 | ||
GenBank mRNA | AK052956.1 | - | 4 | 553 | ||
AK086819.1 | - | 4 | 552 | |||
BC048933.1 | - | 4 | 518 | |||
BC058791.1 | - | 4 | 537 | 6391535 |
