siRNA Details
Gene Symbol: Olfr684 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207249.1 | NP_997132.1 | 1 | 584 | ||
GenBank mRNA | BC120636.1 | - | 1 | 623 | 40129559 | |
BC120638.1 | - | 1 | 623 | 40129561 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership: |
![]() |
Gene Ontology Categories:
