siRNA Details
Gene Symbol: Mcph1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173189.2 | NP_775281.2 | 8 | 1817 | ||
GenBank mRNA | AK042026.1 | - | 7 | 1711 | ||
AK083572.1 | - | 8 | 1860 | |||
AK147997.1 | - | 8 | 1409 | |||
AK148021.1 | - | 8 | 1415 | |||
AY070216.1 | - | 8 | 1397 | |||
BC056924.1 | - | 8 | 1817 | 5697978 |
