siRNA Details
Gene Symbol: Sgcz | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145841.2 | NP_665840.2 | 3, 4 | 1089 | ||
GenBank mRNA | AK048960.1 | - | 3, 4 | 1053 | ||
AK136779.1 | - | 3, 4 | 1090 | |||
AY095374.1 | - | 2, 3 | 305 | |||
AY250713.1 | - | 2, 3 | 305 | |||
BC104048.1 | - | 2, 3 | 305 | 40040143 | ||
BC104049.2 | - | 2, 3 | 305 | 40040144 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
