siRNA Details
Gene Symbol: Adam29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175939.3 | NP_787953.2 | 2 | 889 | ||
GenBank mRNA | AY190759.1 | - | 2 | 873 | ||
BC048676.1 | - | 2 | 887 | |||
BC119089.1 | - | 2 | 870 | 8733838 | ||
BC119091.1 | - | 2 | 870 | 8733840 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
