siRNA Details
Gene Symbol: Podnl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC063764.1 | - | 7 | 842 | 30288186 | |
DQ303463.1 | - | 7 | 827 |

More Information
Other Species Targeted


UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Podnl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC063764.1 | - | 7 | 842 | 30288186 | |
DQ303463.1 | - | 7 | 827 |