siRNA Details
Gene Symbol: Pskh1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173432.2 | NP_775608.1 | 2 | 942 | ||
GenBank mRNA | AK053397.1 | - | 2 | 976 | ||
AK153861.1 | - | 2 | 989 | |||
AK154570.1 | - | 2 | 972 | |||
AK166338.1 | - | 2 | 966 | |||
AK191303.1 | - | 1 | 36 | |||
BC050128.1 | - | 2 | 942 | 30024345 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
