siRNA Details
Gene Symbol: Phlpp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122594.2 | NP_001116066.2 | 3, 4 | 522 | ||
GenBank mRNA | AK048260.1 | - | 3, 4 | 479 | ||
AK048472.1 | - | 3, 4 | 523 | |||
AK165088.1 | - | 3, 4 | 584 | |||
BC117961.1 | - | 3, 4 | 522 | 40098281 | ||
BC117962.1 | - | 3, 4 | 514 | 40098282 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001109131.
Gene Ontology Categories:
