siRNA Details
Gene Symbol: Klhl31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172925.2 | NP_766513.2 | 2 | 684 | ||
GenBank mRNA | AK053085.1 | - | 2 | 685 | ||
BC118027.1 | - | 2 | 683 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
