siRNA Details
Gene Symbol: Fam198a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199927.1 | NP_001186856.1 | 4 | 1631 | ||
NM_177743.5 | NP_808411.1 | 4 | 1562 | |||
GenBank mRNA | AK050223.1 | - | 4 | 1550 | ||
AK134876.1 | - | 4 | 1617 | |||
AK139038.1 | - | 4 | 1632 | |||
AK158406.1 | - | 4 | 1564 | |||
BC113767.1 | - | 3 | 1411 | 40058404 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
