siRNA Details
Gene Symbol: PRH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC136499.1 | - | 3 | 1431 | 9020488 | |
BX641094.1 | - | 3 | 1444 | |||
BX647155.1 | - | 4 | 1338 | |||
BX648656.1 | - | 3 | 1370 | |||
BX648871.1 | - | 2 | 1413 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
