siRNA Details
Gene Symbol: Gm4980 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195529.1 | NP_001182458.1 | 1 | 1437 | ||
GenBank mRNA | AK032609.1 | - | 1 | 510 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: XM_003748929.
Gene Ontology Categories:
