siRNA Details
Gene Symbol: Dkc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001030307.2 | NP_001025478.1 | 9 | 1114 | ||
GenBank mRNA | AK136418.1 | - | 9 | 899 | ||
AK144139.1 | - | 9 | 902 | |||
AK166105.1 | - | 9 | 890 | |||
AK167973.1 | - | 9 | 865 | |||
BC068171.1 | - | 9 | 870 | |||
BC099966.1 | - | 9 | 954 | 6435181 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
