siRNA Details
Gene Symbol: 4930595M18Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173435.3 | NP_775611.2 | 1 | 445 | ||
GenBank mRNA | AK029824.1 | - | 1 | 446 | ||
BB616016.1 | - | 1 | 446 | |||
BC131910.1 | - | 1 | 441 | 40130187 | ||
BC132591.1 | - | 1 | 441 | 40130868 |
