siRNA Details
Gene Symbol: C77370 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077354.2 | NP_001070822.2 | 4 | 2063 | ||
GenBank mRNA | AK038635.1 | - | 4 | 2091 | ||
AK220439.1 | - | 3 | 1908 | |||
DQ975302.1 | - | 4 | 2063 | |||
DQ975303.1 | - | 4 | 2063 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
