siRNA Details
Gene Symbol: Pcdh11x | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271809.1 | NP_001258738.1 | 3 | 2159 | ||
NM_001271810.1 | NP_001258739.1 | 3 | 2159 | |||
GenBank mRNA | AF383875.1 | - | 1 | 578 | ||
AJ564976.1 | - | 3 | 2226 | |||
BC126930.1 | - | 3 | 2082 | 40086368 | ||
EU854474.1 | - | 1 | 921 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
