siRNA Details
Gene Symbol: Pcdh11x | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271809.1 | NP_001258738.1 | 2 | 833 | ||
NM_001271810.1 | NP_001258739.1 | 2 | 833 | |||
GenBank mRNA | AJ564976.1 | - | 2 | 900 | ||
AK039071.1 | - | 2 | 834 | |||
BC126930.1 | - | 2 | 756 | 40086368 |

More Information
Other Species Targeted

Gene Ontology Categories:
