siRNA Details
Gene Symbol: Gprasp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163015.1 | NP_001156487.1 | 7 | 3073 | ||
NM_001163016.1 | NP_001156488.1 | 7 | 3035 | |||
NM_001163017.1 | NP_001156489.1 | 7 | 3020 | |||
GenBank mRNA | AK030638.1 | - | 7 | 3013 | ||
BC117923.2 | - | 7 | 3039 | 40097504 | ||
BC117924.2 | - | 7 | 2989 | 40097505 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
