siRNA Details
Gene Symbol: Tbc1d8b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081499.2 | NP_001074968.1 | 11 | 1976 | ||
GenBank mRNA | BC147581.1 | - | 11 | 1839 | 9056571 | |
BC151057.1 | - | 11 | 1827 | 9087970 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
