siRNA Details
Gene Symbol: Cnksr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177751.2 | NP_808419.1 | 20 | 2743 | ||
GenBank mRNA | AK122399.1 | - | 19 | 2667 | ||
BC043093.1 | - | 20 | 2743 | 6405389 | ||
BC060716.1 | - | 20 | 2743 | 6405389 |
