siRNA Details
Gene Symbol: Tceanc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007577.2 | NP_001007578.2 | 5 | 896 | ||
GenBank mRNA | AK041519.1 | - | 4 | 664 | ||
AK140929.1 | - | 5 | 886 | |||
BC067008.1 | - | 5 | 896 |
