siRNA Details
Gene Symbol: Ccdc15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081429.1 | NP_001074898.1 | 3 | 561 | ||
GenBank mRNA | AK035850.1 | - | 3 | 264 | ||
AK041803.1 | - | 3 | 320 | |||
AK045055.1 | - | 3 | 550 | |||
AK051119.1 | - | 3 | 527 | |||
AK133297.1 | - | 3 | 527 | |||
AK138231.1 | - | 3 | 553 | |||
AK164955.1 | - | 3 | 527 | |||
BC032924.1 | - | 3 | 323 |
