siRNA Details
Gene Symbol: Ccdc15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081429.1 | NP_001074898.1 | 4 | 829 | ||
GenBank mRNA | AK035850.1 | - | 4 | 532 | ||
AK041803.1 | - | 4 | 588 | |||
AK045055.1 | - | 4 | 818 | |||
AK045339.1 | - | 3 | 668 | |||
AK051119.1 | - | 4 | 795 | |||
AK133297.1 | - | 4 | 795 | |||
AK138231.1 | - | 4 | 821 | |||
BC032924.1 | - | 4 | 591 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
