siRNA Details
Gene Symbol: Otoa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139310.1 | NP_647471.1 | 10 | 907 | ||
GenBank mRNA | AK157904.1 | - | 10 | 939 | ||
AK158092.1 | - | 10 | 1022 | |||
AY055122.1 | - | 10 | 907 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
