siRNA Details
Gene Symbol: Mpst | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001162492.1 | NP_001155964.1 | 3 | 718 | ||
NM_001162493.1 | NP_001155965.1 | 3 | 688 | |||
NM_138670.3 | NP_619611.3 | 3 | 822 | |||
GenBank mRNA | AK136571.1 | - | 2 | 717 | ||
BC004079.1 | - | 2 | 663 | 3592659 | ||
BC006066.1 | - | 2 | 663 | |||
BC094469.1 | - | 3 | 822 | 5009481 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
