siRNA Details
Gene Symbol: Vwa1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_147776.4 | NP_680085.3 | 3 | 906 | ||
GenBank mRNA | AK030019.1 | - | 3 | 860 | ||
AK039219.1 | - | 1 | 30 | |||
AK077240.1 | - | 3 | 877 | |||
AY030094.1 | - | 3 | 876 | |||
BC020136.1 | - | 1 | 214 | |||
BC026919.1 | - | 3 | 865 | 3982749 | ||
BC036166.1 | - | 3 | 871 | 4985003 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
