siRNA Details
Gene Symbol: Fcgr4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144559.2 | NP_653142.2 | 3 | 229 | ||
GenBank mRNA | AF499613.1 | - | 3 | 197 | ||
AK076532.1 | - | 3 | 235 | |||
AK087863.1 | - | 3 | 229 | |||
BC027310.1 | - | 3 | 216 | 3674499 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
