siRNA Details
Gene Symbol: AY074887 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145229.2 | NP_660264.1 | 1 | 411 | ||
GenBank mRNA | AY074887.2 | - | 1 | 495 | ||
BC119353.1 | - | 1 | 411 | 8734102 | ||
BC120606.1 | - | 1 | 411 | 40129529 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
