siRNA Details
Gene Symbol: Obox6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145710.2 | NP_663756.2 | 2 | 308 | ||
GenBank mRNA | BC128024.1 | - | 1 | 247 | 40136487 | |
BY731792.1 | - | 2 | 307 | |||
CN712286.1 | - | 2 | 308 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
