siRNA Details
Gene Symbol: Usp7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003918.2 | NP_001003918.2 | 10 | 1082 | ||
GenBank mRNA | AF548565.1 | - | 10 | 1025 | ||
AK135814.1 | - | 1274 | ||||
BC046963.1 | - | 10 | 1020 | |||
BC100666.1 | - | 5 | 388 |

More Information
Other Species Targeted

Gene Ontology Categories:
