siRNA Details
Gene Symbol: Vmn1r69 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145842.3 | NP_665841.1 | 1 | 1212 | ||
GenBank mRNA | AF394954.1 | - | 1 | 1211 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
