siRNA Details
Gene Symbol: Cables2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145851.2 | NP_665850.2 | 7, 8 | 995 | ||
GenBank mRNA | AK050121.1 | - | 4, 5 | 2617 | ||
BC036954.1 | - | 2, 3 | 386 | |||
BC037028.1 | - | 2, 3 | 386 | |||
BC052789.1 | - | 1, 2 | 337 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
