siRNA Details
Gene Symbol: Ropn1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145852.2 | NP_665851.2 | 4 | 715 | ||
GenBank mRNA | AF305427.1 | - | 4 | 672 | ||
AK131675.1 | - | 4 | 764 | |||
BC048680.1 | - | 4 | 651 | 6774441 | ||
BC060990.1 | - | 4 | 659 | 6774094 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
