siRNA Details
Gene Symbol: Nod2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145857.2 | NP_665856.2 | 4 | 1720 | ||
GenBank mRNA | AF520774.1 | - | 4 | 1765 | ||
AK089390.1 | - | 4 | 1684 | |||
AK089843.1 | - | 4 | 1684 | |||
AY160220.1 | - | 3 | 1649 | |||
AY160221.1 | - | 4 | 1721 | |||
BC044774.2 | - | 4 | 1747 |
