siRNA Details
Gene Symbol: Olfr1116 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011734.1 | NP_001011734.1 | 1 | 90 |
Gene Symbol: Olfr1118 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207632.2 | NP_997515.2 | 1 | 117 | ||
GenBank mRNA | BC120792.1 | - | 1 | 117 | 40129715 |
