siRNA Details
Gene Symbol: Olfr317 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011769.2 | NP_001011769.2 | 1 | 765 | ||
GenBank mRNA | BC147131.1 | - | 1 | 765 | 9056194 | |
BC147132.1 | - | 1 | 765 | 9056174 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
