siRNA Details
Gene Symbol: Olfr285 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011778.1 | NP_001011778.1 | 1 | 387 |
Gene Symbol: Olfr257 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207554.1 | NP_997437.1 | 1 | 387 |
