siRNA Details
Gene Symbol: Olfr1039 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011784.2 | NP_001011784.2 | 1 | 135 |
Gene Symbol: Olfr1040 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207561.2 | NP_997444.2 | 1 | 135 |
