siRNA Details
Gene Symbol: Olfr1251 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011529.1 | NP_001011529.1 | 1 | 790 | ||
GenBank mRNA | BC147159.1 | - | 1 | 846 | 9056474 | |
BC147160.1 | - | 1 | 890 | 9056173 |
Gene Symbol: Olfr1249 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011796.2 | NP_001011796.2 | 1 | 790 |
Gene Symbol: Olfr1253 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146373.1 | NP_666485.1 | 1 | 790 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
