siRNA Details
Gene Symbol: Olfr1306 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011803.2 | NP_001011803.2 | 1 | 838 |
Gene Symbol: Olfr1307 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011787.1 | NP_001011787.1 | 1 | 838 |
