siRNA Details
Gene Symbol: Olfr1385 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011805.1 | NP_001011805.1 | 1 | 788 |
Gene Symbol: Olfr1386 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011741.2 | NP_001011741.1 | 1 | 887 | ||
GenBank mRNA | BC147372.1 | - | 1 | 887 | 9056436 | |
BC147373.1 | - | 1 | 887 | 9056431 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
