siRNA Details
Gene Symbol: Olfr193 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011791.1 | NP_001011791.1 | 1 | 650 | ||
GenBank mRNA | BC147133.1 | - | 1 | 675 | 9056183 | |
BC147134.1 | - | 1 | 673 | 9056182 |
Gene Symbol: Olfr191 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011807.2 | NP_001011807.2 | 1 | 650 |
