siRNA Details
Gene Symbol: Olfr93 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011813.1 | NP_001011813.1 | 1 | 670 | ||
GenBank mRNA | BC139238.1 | - | 1 | 751 | 8862260 | |
BC139240.1 | - | 1 | 751 | 8862262 |
