siRNA Details
Gene Symbol: Olfr316 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011818.2 | NP_001011818.2 | 1 | 517 | ||
GenBank mRNA | BC150858.1 | - | 1 | 580 | 9087769 |
Gene Symbol: Olfr318 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146501.2 | NP_666712.2 | 1 | 565 | ||
GenBank mRNA | BC137758.1 | - | 1 | 565 | 8860777 |
Gene Symbol: Olfr319 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC104265.1 | - | 1 | 517 | 40040340 |
Gene Symbol: Olfr320 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207230.1 | NP_997113.1 | 1 | 517 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
