siRNA Details
Gene Symbol: Olfr1330 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146334.2 | NP_666446.1 | 1 | 352 | ||
GenBank mRNA | BC108943.1 | - | 1 | 352 | 40042936 | |
BC108944.1 | - | 1 | 347 |
Gene Symbol: Olfr1329 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011870.2 | NP_001011870.2 | 1 | 346 |
Gene Symbol: Olfr1331 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011856.2 | NP_001011856.2 | 1 | 346 |
Gene Symbol: Olfr1337 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146309.3 | NP_666421.2 | 1 | 355 | ||
GenBank mRNA | BC127989.1 | - | 1 | 351 | 40135644 | |
BC152318.1 | - | 1 | 357 |

More Information
Other Species Targeted

Gene Ontology Categories:
