siRNA Details
Gene Symbol: Olfr685 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011857.1 | NP_001011857.1 | 1 | 592 | ||
GenBank mRNA | BC147196.1 | - | 1 | 681 | 9056252 | |
BC147197.1 | - | 1 | 681 | 9056258 |

More Information
Other Species Targeted

Gene Ontology Categories:
